Thermo Scientific Chemicals

2,4,6-Triiodophenol, 98%, Thermo Scientific Chemicals

Catalog number: A17145.22
100 g, Each
Thermo Scientific Chemicals

2,4,6-Triiodophenol, 98%, Thermo Scientific Chemicals

Catalog number: A17145.22
100 g, Each
Quantity
Catalog number: A17145.22
also known as A17145-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
609-23-4
IUPAC Name
2,4,6-triiodophenol
Molecular Formula
C6H3I3O
InChI Key
VAPDZNUFNKUROY-UHFFFAOYSA-N
SMILES
OC1=C(I)C=C(I)C=C1I
Appearance (Color)
White to dark cream to yellow to brown or pale pink
Form
Powder
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

Solubility
Insoluble in water.

Notes
Incompatible withacid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text