Thermo Scientific Chemicals

N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: A17164.09
10 g, Each
Thermo Scientific Chemicals

N-(1-Naphthyl)ethylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: A17164.09
10 g, Each
Quantity
Catalog number: A17164.09
also known as A17164-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1465-25-4
IUPAC Name
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular Formula
C12H16Cl2N2
InChI Key
MZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
Appearance (Color)
White to cream or pale grey
Form
powder
Assay (Titration ex Chloride)
> 95.0% (dry wt. basis)
Water Content (Karl Fischer Titration)
< 5.0%

Description

N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

Solubility
Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.

Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text