2-Chloro-6-nitrophenol, 98%, Thermo Scientific Chemicals
2-Chloro-6-nitrophenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-6-nitrophenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A17172.06
also known as A17172-06
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
2-Chloro-6-nitrophenol, 98%, Thermo Scientific Chemicals
Catalog numberA17172.06
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS603-86-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)65.5-71.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only