Thermo Scientific Chemicals

(S)-(-)-1-(1-Naphthyl)ethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A17176.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-(1-Naphthyl)ethylamine, 99%, Thermo Scientific Chemicals

Catalog number: A17176.06
5 g, Each
Quantity
Catalog number: A17176.06
also known as A17176-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10420-89-0
IUPAC Name
(1S)-1-(naphthalen-1-yl)ethan-1-aminium
Molecular Formula
C12H14N
InChI Key
RTCUCQWIICFPOD-VIFPVBQESA-O
SMILES
C[C@H]([NH3+])C1=C2C=CC=CC2=CC=C1
Form
Liquid
Assay from Supplier's CofA
≥98.5% (US-sourced material)
Comment
May darken on storage
Refractive Index
1.6215-1.6245 @ 20?C (UK-sourced material)
Comment
Material sourced in UK and US

Description

(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(1-Naphthyl)ethylamine, (S)-(-)-1-(1-Naphthyl)ethylamine is used in the asymmetric synthesis of α-cyanocarboxylates. Also used in the synthesis of chiral imadazolin-2-ylidene ligands used in organometallic catalysis. Chiral/Asymmetric synthesis.

Solubility
Soluble in chloroform, ethanol.

Notes
Air Sensitive. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text