Thermo Scientific Chemicals

2,6-Dibromo-4-methylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A17184.22
100 g, Each
Thermo Scientific Chemicals

2,6-Dibromo-4-methylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A17184.22
100 g, Each
Quantity
Catalog number: A17184.22
also known as A17184-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6968-24-7
IUPAC Name
2,6-dibromo-4-methylaniline
Molecular Formula
C7H7Br2N
InChI Key
ATDIROHVRVQMRO-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=C(N)C(Br)=C1
Appearance (Color)
Cream
Melting Point (clear melt)
70.5-76.5?C
Assay (GC)
≥98.0%
Form
Powder

Description

2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromo-4-methylaniline is used as coupling reagent in spectrophotometric determination of carbaryl in various environmental samples. It was also in preparation of 2-bromo-6-iodo-4-methylaniline. 2,6-Dibromo-4-methylaniline undergoes Suzuki-coupling reaction with 2-dihydroxyboryl-3-methyl-2-cyclopenten-1-one during synthesis of the dinuclear dichlorotitanium complexes. 2,6-Dibromo-4-methylaniline is also used as an intermediate for the synthesis of pharmaceutucals, agrochemicals, dyes and other organic chemicals.

Solubility
Insoluble in water.

Notes
Stable under ordinary conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text