Thermo Scientific Chemicals

Tetra-n-butylammonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17196.36
500 g, Each
Thermo Scientific Chemicals

Tetra-n-butylammonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17196.36
500 g, Each
Quantity
Catalog number: A17196.36
also known as A17196-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3109-63-5
IUPAC Name
hexafluoro-λ⁵-phosphanuide; tetrabutylazanium
Molecular Formula
C16H36F6NP
InChI Key
BKBKEFQIOUYLBC-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Form
Crystals or powder or crystalline powder
Elemental Analysis
Hydrogen H: 9.13-9.60% (Theory: 9.36(5)%)
Elemental Analysis
Si: <1000ppm
Elemental Analysis
Carbon C: 48.36-50.84% (Theory: 49.60%)
Elemental Analysis
Nitrogen N: 3.53-3.71% (Theory: 3.61(5)%)

Description

Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

Solubility
Soluble in acetone, acetonitrile and tetrhydrofuran.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text