2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17201.06
also known as A17201-06
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals
Catalog numberA17201.06
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3682-35-7
IUPAC Nametris(pyridin-2-yl)-1,3,5-triazine
Molecular FormulaC18H12N6
InChI KeyKMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)247.0-253.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (HPLC)≥97.5%
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
RUO – Research Use Only

General References:

  1. Reagent for determination of Fe: Anal. Chem., 31, 1862 (1959).
  2. Gaba, M.; Mohan, C. Design, Synthesis and Biological Evaluation of Novel 1,2,5-Substituted Benzimidazole Derivatives as Gastroprotective Anti-inflammatory and Analgesic Agents. Med. Chem. 2015, 5 (2), 058-063.
  3. Zhang, Y. Z.; Zhao, H. H.; Funck, E.; Dunbar, K. R. A Single-Chain Magnet Tape Based on Hexacyanomanganate(III). Angew. Chem. Int. Ed. 2015, 127 (19), 5675-5679.