Thermo Scientific Chemicals

2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Catalog number: A17201.22
100 g, Each
Thermo Scientific Chemicals

2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Catalog number: A17201.22
100 g, Each
Quantity
Catalog number: A17201.22
also known as A17201-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3682-35-7
IUPAC Name
tris(pyridin-2-yl)-1,3,5-triazine
Molecular Formula
C18H12N6
InChI Key
KMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILES
C1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
Melting Point (clear melt)
247.0-253.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%

Description

Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the colorimetric determination of iron2,4,6-Tri(2-pyridyl)-1,3,5-triazine is used in the spectrophotometric determination of iron. It acts as a tridendate ligand and form coordination complexes with lanthanides and rhodium.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text