Used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a chemical intermediate.
Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
- M B Lis.; V V Kazakova.; N A Martynova.; V P Rogachkov.; V V Sotnik. Establishment of the maximum permissible concentration of 2-brompentane in the air of a work area.Gig Tr Prof Zabol.1981,5 51-52.
- S Sekiguchi.; T Honma. Industrial Health 1998-07-01 Influence of 2-bromopropane on reproductive system--2-bromopropane inhibits forced ovulation in mice.Ind Health.1998,36,(3), 297-299.