Thermo Scientific Chemicals

Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals

Catalog number: A17213.30
250 g, Each
Thermo Scientific Chemicals

Hexamethylenetetramine, 99+%, Thermo Scientific Chemicals

Catalog number: A17213.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17213.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-97-0
IUPAC Name
1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular Formula
C6H12N4
InChI Key
VKYKSIONXSXAKP-UHFFFAOYSA-N
SMILES
C1N2CN3CN1CN(C2)C3
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥99.0 to ≤101.0%
Water Content (Karl Fischer Titration)
≤1%
Form
Crystals or powder or crystalline powder or chunks or fused solid
Identification (FTIR)
Conforms

Description

Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylenetetramine is used as a formylation agent in the synthesis and as a precipitating agent. It is also used in the production of phenolic resins and its moulding compounds, which finds application as binders in fire proof materials, brake and clutch linings. It is involved in the preparation of 5-tert-butyl-2-hydroxy-isophthalaldehyde. It is useful for the treatment of urinary tract infection. Further, it is used in the Duff reaction, Sommelet reaction and in the Delepine reaction.

Solubility
Soluble in water, ethanol, chloroform and acetone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids. Avoid conditions like flames, sparks and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text