Thermo Scientific Chemicals

1-(2-Hydroxyethyl)piperazine, 98+%, Thermo Scientific Chemicals

Catalog number: A17215.09
10 g, Each
Thermo Scientific Chemicals

1-(2-Hydroxyethyl)piperazine, 98+%, Thermo Scientific Chemicals

Catalog number: A17215.09
10 g, Each
Quantity
Catalog number: A17215.09
also known as A17215-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-76-4
IUPAC Name
2-(piperazin-1-yl)ethan-1-ol
Molecular Formula
C6H14N2O
InChI Key
WFCSWCVEJLETKA-UHFFFAOYSA-N
SMILES
OCCN1CCNCC1
Appearance (Color)
Clear colorless
Assay (Silylated GC)
≥98.0%
Form
Liquid or viscous liquid
Refractive Index
1.5045-1.5085 @ 20?C

Description

1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.

Solubility
Miscible with water. Also miscible in ethanol, acetone, ether benzene. Immiscible in hexane.

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text