1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Piperazineethanol is a hydroxyalkyl substituted pieprazine used in the preparation of pharmaceutical compounds. It is also used as an Intermediate in the manufacture of polyurethane catalysts, corrosion inhibitors, surfactants, synthetic fibers, pharmaceuticals. It is also used to synthesize triethylenediamine.
Solubility
Miscible with water. Also miscible in ethanol, acetone, ether benzene. Immiscible in hexane.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
- I Sircar.; SJ Haleen.; SE Burke. Synthesis and biological activity of 4-(diphenylmethyl)-. alpha.-[(4-quinolinyloxy) methyl]-1-piperazineethanol and related compounds. J. Med. Chem..199235 (23), 4442-4449.
- HL Yale. Esters of 4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]-1-piperazineethanol and related compounds as long-acting antipsychotic agents. Synthesis of the 1-adamantoate, the first crystalline base. 4 J. Med. Chem.197720 (2), 302-304.