Thermo Scientific Chemicals

3,3'-Thiodipropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17220.30
250 g, Each
Thermo Scientific Chemicals

3,3'-Thiodipropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: A17220.30
250 g, Each
Quantity
Catalog number: A17220.30
also known as A17220-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-17-1
IUPAC Name
3-[(2-carboxyethyl)sulfanyl]propanoic acid
Molecular Formula
C6H10O4S
InChI Key
ODJQKYXPKWQWNK-UHFFFAOYSA-N
SMILES
OC(=O)CCSCCC(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5% to ≤102.5% (Dry wt. basis)

Description

3,3'-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

Solubility
Soluble in acetone, ethanol. Slightly soluble in water.

Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text