N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
Solubility
It has optimal solubility in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
- M Kraszni.; I Bányai.; B Noszál. Determination of conformer-specific partition coefficients in octanol/water systems. Journal of medicinal chemistry. 200346 (11), 2241-2245.
- SNG Tyler.; RL Webster. Sterically hindered malonamide monomers for the step growth synthesis of polyesters and polyamides. Chemical Communications. 201450 (1), 10665-10668.