Isopropyl formate, 98%, Thermo Scientific Chemicals
Isopropyl formate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl formate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A17222.30
also known as A17222-30
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
Isopropyl formate, 98%, Thermo Scientific Chemicals
Catalog numberA17222.30
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
CAS625-55-8
IUPAC Namepropan-2-yl formate
Molecular FormulaC4H8O2
InChI KeyRMOUBSOVHSONPZ-UHFFFAOYSA-N
SMILESCC(C)OC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.3660-1.3700 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

Solubility
slightly soluble in water, completely miscible with alcohol, ether, and most organic solvents.

Notes
d^2^0 0.901. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. JE Piercy.; SV Subrahmanyam. Ultrasonic Relaxation and Cis—Trans Isomerization in Isopropyl Formate. The Journal of Chemical Physics. 196542 (1), 1475 -1477.
  2. P Simmons.; CK Fisher. Ethyl formate and isopropyl formate as fumigants for packages of dried fruits. Journal of Economic Entomology. 194538 (6), 715 -716.