Thermo Scientific Chemicals

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, Thermo Scientific Chemicals

Catalog number: A17225.06
5 g, Each
Thermo Scientific Chemicals

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%, Thermo Scientific Chemicals

Catalog number: A17225.06
5 g, Each
Quantity
Catalog number: A17225.06
also known as A17225-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1450-63-1
IUPAC Name
(1,4,4-triphenylbuta-1,3-dien-1-yl)benzene
Molecular Formula
C28H22
InChI Key
KLCLIOISYBHYDZ-UHFFFAOYSA-N
SMILES
C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
195.0-204.0?C

Description

1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

Solubility
Soluble in ethanol, benzene, chloroform, tolune and acetic acid.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text