Thermo Scientific Chemicals

L-Glutamic acid monopotassium salt monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: A17232.0B
1000 g, Each
Thermo Scientific Chemicals

L-Glutamic acid monopotassium salt monohydrate, 97+%, Thermo Scientific Chemicals

Catalog number: A17232.0B
1000 g, Each
Quantity
Catalog number: A17232.0B
also known as A17232-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6382-01-0
IUPAC Name
potassium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular Formula
C5H10KNO5
InChI Key
XIBUKSQTWSKJMQ-QTNFYWBSSA-M
SMILES
O.[K+].N[C@@H](CCC([O-])=O)C(O)=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.0 to ≤103.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
4.64-12.73% (0.5-1.5 waters)
Optical Rotation
22.5 ± 1.0? (C=1 in 5N HCl)

Description

L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text