Thermo Scientific Chemicals

2,6-Di-tert-butylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A17244.0I
5000 g, Each
Thermo Scientific Chemicals

2,6-Di-tert-butylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A17244.0I
5000 g, Each
Quantity
Catalog number: A17244.0I
also known as A17244-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128-39-2
IUPAC Name
2,6-di-tert-butylphenol
Molecular Formula
C14H22O
InChI Key
DKCPKDPYUFEZCP-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
Appearance (Color)
White to cream to yellow or pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
34-39?C
Form
Crystalline or fused solid

Description

Widely used as antioxidant in fuels, lubricants and polymers. Employed as a synthetic intermediate for the production of higher molecular weight phenolic antioxidants. Used as an oxidation inhibitor and stabilizer (e.g. for fuel, oil and gasoline) and also used in plastics and rubber. Also applied as an intermediate and an antioxidant in aviation gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used as antioxidant in fuels, lubricants and polymers. Employed as a synthetic intermediate for the production of higher molecular weight phenolic antioxidants. Used as an oxidation inhibitor and stabilizer (e.g. for fuel, oil and gasoline) and also used in plastics and rubber. Also applied as an intermediate and an antioxidant in aviation gasoline.

Solubility
Soluble in water at 20°C 0.003g/L. Solubility in methanol is almost transparent.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text