Thermo Scientific Chemicals

Aliquat|r 336 TG, Thermo Scientific Chemicals

Catalog number: A17247.AP
500 mL, Each
Thermo Scientific Chemicals

Aliquat|r 336 TG, Thermo Scientific Chemicals

Catalog number: A17247.AP
500 mL, Each
Quantity
Catalog number: A17247.AP
also known as A17247-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63393-96-4
IUPAC Name
methyltrioctylazanium chloride
Molecular Formula
C25H54ClN
InChI Key
XKBGEWXEAPTVCK-UHFFFAOYSA-M
SMILES
[Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
Appearance (Color)
Clear pale yellow
Form
Viscous liquid
Assay from Supplier's CofA
112-118 (Quaternary value)
Assay from Supplier's CofA
88.2-93% quaternary ammonium salts
Comment
Isomer content (C8): ca 60% (balance other chain lengths)

Description

Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

Solubility
Miscible with benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text