It is the intermediate in the synthesis of potent antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is the intermediate in the synthesis of potent antitumor agents.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.
Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
RUO – Research Use Only
Zhenrong Guo.; Eric D Dowdy.; Wen-Sen Li.; Richard Polniaszek.; Edward Delaney. A novel method for the mild and selective amidation of diesters and the amidation of monoesters. Tetrahedron Lett. 2001, 42 (10),1843-1845.
Richard D. Webster.; Alan M. Bond.; Richard G. Compton. Voltammetric and EPR Spectroscopic Studies Associated with the Reduction of Pyridine- and Benzene-Substituted n-Alkyl Esters and Thioic S-Esters in Aprotic Solvents. J. Phys. Chem. 1996, 100 (24),10288-10297.