Thermo Scientific Chemicals

Dimethyl pyridine-2,5-dicarboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: A17250.22
100 g, Each
Thermo Scientific Chemicals

Dimethyl pyridine-2,5-dicarboxylate, 98+%, Thermo Scientific Chemicals

Catalog number: A17250.22
100 g, Each
Quantity
Catalog number: A17250.22
also known as A17250-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
881-86-7
IUPAC Name
2,5-dimethyl pyridine-2,5-dicarboxylate
Molecular Formula
C9H9NO4
InChI Key
TUGSJNQAIMFEDY-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(N=C1)C(=O)OC
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
160.0-167.0°C

Description

It is the intermediate in the synthesis of potent antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the intermediate in the synthesis of potent antitumor agents.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents, reducing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text