Anthracene, 97%, Thermo Scientific Chemicals
Anthracene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Anthracene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17261.0I
also known as A17261-0I
Price (USD)/ Each
1,217.00
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
1,217.00
Add to cart
Anthracene, 97%, Thermo Scientific Chemicals
Catalog numberA17261.0I
Price (USD)/ Each
1,217.00
-
Add to cart
Chemical Identifiers
CAS120-12-7
IUPAC Nameanthracene
Molecular FormulaC14H10
InChI KeyMWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILESC1=CC2=CC3=CC=CC=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to green to pale brown to black
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)213.0-222.0?C
Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used as a source of dyestuffs and involved in the preparation of red dye alizarin. It is an organic semiconductor used as a scintillator for detectors of high energy photons, electrons and alfa particles. It is doped with polyvinyltoluene to prepare plastic scintillator. Further, it is used in the preparation of anthraquinone, wood preservatives, coating materials and pigments. In addition to this, it is used to prepare 1-hydroxyanthracene, 2-hydroxyanthracene and in 9,10-dihydroxyanthracene.

Solubility
Soluble in methanol, benzene, chloroform, ether, carbon disulfide, carbon tetrachloride and toluene. Slightly soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and hypochlorites.
RUO – Research Use Only

General References:

  1. Malkondu, S.; Turhan, D.; Kocak, A. Copper(II)-directed static excimer formation of an anthracene-based highly selective fluorescent receptor. Tetrahedron Lett. 2015, 56 (1), 162-167.
  2. Ma, Z.; Yang, F.; Wang, Z.; Jia, X. Mechanically induced color change based on the chromophores of anthracene and rhodamine 6G. Tetrahedron Lett. 2015, 56 (2), 393-396.