Azelaic acid, tech. 80%, Thermo Scientific Chemicals
Azelaic acid, tech. 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Azelaic acid, tech. 80%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A17271.36
also known as A17271-36
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
Add to cart
Azelaic acid, tech. 80%, Thermo Scientific Chemicals
Catalog numberA17271.36
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
-
Add to cart
Chemical Identifiers
CAS123-99-9
IUPAC Namenonanedioic acid
Molecular FormulaC9H16O4
InChI KeyBDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Formcrystalline flakes or powder
Assay (Silylated GC)≥75.0%
Assay (Aqueous acid-base Titration)≥97.0% (total isomers)
Identification (FTIR)Conforms
It is applied in organic synthesis, lacquers, polyester adhesives, and urethane elastomers. Azelaic acid directly inhibits kallikrein 5 (KLK5), down regulating expression of KLK5, toll-like receptor 2 (TLR 2), and cathelicidin, and suppresses activity of serine proteases. In PUFA-treated fibroblasts, azelaic acid decreases generation of ROS and up regulates expression of antioxidative enzymes, inhibiting cell damage; agonist activity at PPAR? is thought to drive these effects. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. M.R. Gasco, et al. In vitro permeation of azelaic acid from viscosized microemulsions.Int. J. Pharm.,1991,69(3), 193-196.
  2. Graupe K, et al. Efficacy and safety of topical azelaic acid (20 percent cream): an overview of results from European clinical trials and experimental reports.European Pubmed.,1996,57 (1), 20-35.