Thermo Scientific Chemicals

2,4,5-Triphenylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: A17291.09
10 g, Each
Thermo Scientific Chemicals

2,4,5-Triphenylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: A17291.09
10 g, Each
Quantity
Catalog number: A17291.09
also known as A17291-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
484-47-9
IUPAC Name
2,4,5-triphenyl-1H-imidazole
Molecular Formula
C21H16N2
InChI Key
RNIPJYFZGXJSDD-UHFFFAOYSA-N
SMILES
N1C(=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Melting Point (clear melt)
271.5-280.5°C
Appearance (Color)
White to off-white to pale yellow
Assay (unspecified)
≥96.0 to ≤104.0%

Description

2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text