Thermo Scientific Chemicals

N-Acetyl-L-tyrosine, 99%

Catalog number: A17307.18
50 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-tyrosine, 99%

Catalog number: A17307.18
50 g, Each
Quantity
Catalog number: A17307.18
also known as A17307-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
537-55-3
IUPAC Name
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C11H13NO4
InChI Key
CAHKINHBCWCHCF-UEQNJFAPNA-N
SMILES
CC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Melting Point (clear melt)
149.0-155.0°C
Optical Rotation
+47° to +49° (c=1 in water)
Form
Crystals or powder or crystalline powder

Description

N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water (25 mg/ml), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text