N-Acetyl-L-tyrosine, 99%
N-Acetyl-L-tyrosine, 99%
Thermo Scientific Chemicals

N-Acetyl-L-tyrosine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A17307.30
also known as A17307-30
Price (USD)/ Each
648.65
Online exclusive
721.00 
Save 72.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
648.65
Online exclusive
721.00 
Save 72.35 (10%)
Add to cart
N-Acetyl-L-tyrosine, 99%
Catalog numberA17307.30
Price (USD)/ Each
648.65
Online exclusive
721.00 
Save 72.35 (10%)
-
Add to cart
Chemical Identifiers
CAS537-55-3
IUPAC Name(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC11H13NO4
InChI KeyCAHKINHBCWCHCF-UEQNJFAPNA-N
SMILESCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)149.0-155.0°C
Optical Rotation+47° to +49° (c=1 in water)
FormCrystals or powder or crystalline powder
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water (25 mg/ml), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Harold Edelhoch. Spectroscopic Determination of Tryptophan and Tyrosine in Proteins. Biochemistry. 1967, 6 (7),1948-1954.
  2. Inger Magnusson.; Lars Ekman.; Marie Wångdahl.; John Wahren. N-acetyl-l-tyrosine and N-acetyl-l-cysteine as tyrosine and cysteine precursors during intravenous infusion in humans.Metabolism. 1989, 38 (10),957-961.