Triethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Triethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number A17318.0B
also known as A17318-0B
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
Add to cart
Triethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberA17318.0B
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS554-68-7
IUPAC Namehydrogen triethylamine chloride
Molecular FormulaC6H16ClN
InChI KeyILWRPSCZWQJDMK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (on dried substance)
Loss on Drying≤1.0%
Appearance (Color)White to cream
It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a protein purification reagent. Triethylamine is used as a competing base for the separation of acidic basic and neutral drugs by reverse-phased high-performance liquid chromatography (HPLC). Triethylamine induces visual disturbances (such as foggy vision) in humans, and is also used in industry as a quenching agent in the ozonolysis of alkenes (e.g. (E)-2-Pentene.

Solubility
Soluble in water at 20°C 1440 g/L. Soluble in ethanol, chloroform, and alcohol. Insoluble in ether.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only

General References:

  1. James R. Vaughan, et al. The Preparation of Peptides Using Mixed Carbonic—Carboxylic Acid Anhydrides.J. Am. Chem. Soc.,1952,74(3), 676-678.
  2. P. W. Morgan. Synthesis and Properties of Aromatic and Extended Chain Polyamides.Macromolecules.,1977,10 (6), 1381-1390.