It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the modification of the photochemical behavior of organic molecules by cyclodextrin.
Solubility
Sparingly soluble (0.061 g/L) (25°C) in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
James H. Brewster, et al. The Configuration of Atrolactic Acid. Retention of Configuration in the Acid-catalyzed Ring Opening of Stilbene Oxides.J. Am. Chem. Soc.,1956,78(16), 4061-4064.
M.S. Syamala. Modification of the photochemical behaviour of organic molecules by cyclodextrin: Geometric isomerization of stilbenes and alkyl cinnamates.J. Photochem.,1986,34 (2), 219-229.