Thermo Scientific Chemicals

4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A17337.03
1 g, Each
Thermo Scientific Chemicals

4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: A17337.03
1 g, Each
Quantity
Catalog number: A17337.03
also known as A17337-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10130-89-9
IUPAC Name
4-(chlorosulfonyl)benzoic acid
Molecular Formula
C7H5ClO4S
InChI Key
PTCSSXYPZOFISK-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%

Description

It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a chemical reagent used in catalyzing living radical polymerizations, in the preparation of polymer bound transfer hydrogenation catalyst1 and 4-(carboxymethyl-sulfamoyl)-benzoyladenosine. It is also used as a reagent in the sulfonation of γ-cyclodextrin.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text