Methyl Trifluoroacetate, 98+%, Thermo Scientific Chemicals
Methyl Trifluoroacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl Trifluoroacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A17338.18
also known as A17338-18
Price (USD)/ Each
71.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
71.00
Add to cart
Methyl Trifluoroacetate, 98+%, Thermo Scientific Chemicals
Catalog numberA17338.18
Price (USD)/ Each
71.00
-
Add to cart
Chemical Identifiers
CAS431-47-0
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>98.0%
Refractive Index<1.3000 @ 20?C
Appearance (Color)Clear, colorless
Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl­guanidine, has also been used for the N-acylation of amino acids. It is used as an intermediate in the synthesis of pharmaceutical and agricultural chemicals. The chemical reactions usually involve nucleophilic substitution of the ester and are useful for the preparation of trifluoroacylated compounds.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only

General References:

  1. Langlois BR and Roques N. Nucleophilic trifluoromethylation of aryl halides with methyl trifluoroacetate.J. Fluor. Chem.,2007,128(10), 1318-1325.
  2. M. N. Vargaftik, et al. Highly selective partial oxidation of methane to methyl trifluoroacetate.J. Chem. Soc., Chem. Commun.,1990,15, 1049-1050.
  3. Useful reagent for trifluoroacetylation of amines under mild conditions. In the presence of 1,1,3,3-Tetramethyl guanidine, A12314, has also been used for the N-acylation of amino acids: Synthesis, 399 (1976).