Thermo Scientific Chemicals

2-Phenylcyclohexanone, 98%, Thermo Scientific Chemicals

Catalog number: A17340.03
1 g, Each
Thermo Scientific Chemicals

2-Phenylcyclohexanone, 98%, Thermo Scientific Chemicals

Catalog number: A17340.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17340.03
also known as A17340-03
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
1444-65-1
IUPAC Name
2-phenylcyclohexan-1-one
Molecular Formula
C12H14O
InChI Key
DRLVMOAWNVOSPE-UHFFFAOYNA-N
SMILES
O=C1CCCCC1C1=CC=CC=C1
Melting Point (clear melt)
55.0-61.0?C
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
White to cream
Assay (GC)
≥97.5%

Description

It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text