It can be used to produce 6-oxo-6-phenyl-hexanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used to produce 6-oxo-6-phenyl-hexanoic acid.
Solubility
Slightly soluble in water.
Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
Enrique Díez-Barra.; Antonio de la Hoz.; Sonia Merino.; Prado Sánchez-Verdú. Regioselectivity and diastereoselectivity in the phase transfer catalysed Michael addition of 2-phenylcyclohexanone. Tetrahedron Lett. 1997, 38 (13), 2359-2362.
Stephanie D. Meyer.; Stuart L. Schreiber. Acceleration of the Dess-Martin Oxidation by Water. J. Org. Chem. 1994, 59 (24),7549-7552.