Thermo Scientific Chemicals

Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals

Catalog number: A17342.0B
1000 g, Each
Thermo Scientific Chemicals

Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals

Catalog number: A17342.0B
1000 g, Each
Quantity
Catalog number: A17342.0B
also known as A17342-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80-09-1
IUPAC Name
4-(4-hydroxybenzenesulfonyl)phenol
Molecular Formula
C12H10O4S
InChI Key
VPWNQTHUCYMVMZ-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
Assay (HPLC)
≥98.5%
Appearance (Color)
White to pale cream
Melting Point (clear melt)
244.0-251.0?C
Form
Powder

Description

It is a reactant used in epoxy reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant used in epoxy reactions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text