Sodium metabisulfite, 97%, Thermo Scientific Chemicals
Sodium metabisulfite, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium metabisulfite, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
10,000 g
Catalog number A17351.36
Price (USD)/ Each
26.65
Online exclusive
28.90 
Save 2.25 (8%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
28.90 
Save 2.25 (8%)
Add to cart
Sodium metabisulfite, 97%, Thermo Scientific Chemicals
Catalog numberA17351.36
Price (USD)/ Each
26.65
Online exclusive
28.90 
Save 2.25 (8%)
-
Add to cart
Chemical Identifiers
CAS7681-57-4
IUPAC Namedisodium (sulfinatooxy)sulfinate
Molecular FormulaNa2O5S2
InChI KeyLDTLADDKFLAYJA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)OS([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Iodometric Titration)≥96.0 to ≤104.0%
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium metabisulfite is used as an active ingredient in campden tablets, as a cleaning agent for potable water reverse osmosis membranes, as a preservative and an antioxidant in food. It also acts as a corrosion inhibitor in oil and gas industry, as a bleaching agent in the production of coconut cream, as the source of sulfur dioxide and in the destruction of cyanide in commercial gold cyanidation processes. It is utilized in the precipitation of gold from auric acid as well as in the waste water treatment to remove hexavaent chromium as trivalent chromium by precipitation after reduction. Further, it is used in photography. In addition, it plays as an oxygen scavenger to eliminate the dissolved oxygen in wastewater and in pipes.

Solubility
Soluble in water, ethanol and glycerol.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Tan, T. C.; Cheng, L. H.; Bhat, R.; Rusul, G.; Easa, A. M. Effectiveness of ascorbic acid and sodium metabisulfite as anti-browning agent and antioxidant on green coconut water (Cocos nucifera) subjected to elevated thermal processing. Int. Food Res. J. 2015, 22 (2), 631-637
  2. Zaid, B.; Maddache, N.; Saidi, D.; Souami, N.; Bacha, N.; Ahmed, A. S. Electrochemical evaluation of sodium metabisulfite as environmentally friendly inhibitor for corrosion of aluminum alloy 6061 in a chloride solution. J. Alloys Compd. 2015, 629, 188-196.