Thermo Scientific Chemicals

2-Ethylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A17352.36
500 g, Each
Thermo Scientific Chemicals

2-Ethylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A17352.36
500 g, Each
Quantity
Catalog number: A17352.36
also known as A17352-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-71-0
IUPAC Name
2-ethylpyridine
Molecular Formula
C7H9N
InChI Key
NRGGMCIBEHEAIL-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=N1
Refractive Index
1.4955-1.4995 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.

Solubility
Miscible with water (42 g/L at20°C).

Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text