Thermo Scientific Chemicals

8-Aminooctanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A17353.03
1 g, Each
Thermo Scientific Chemicals

8-Aminooctanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A17353.03
1 g, Each
Quantity
Catalog number: A17353.03
also known as A17353-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1002-57-9
IUPAC Name
8-aminooctanoic acid
Molecular Formula
C8H17NO2
InChI Key
UQXNEWQGGVUVQA-UHFFFAOYSA-N
SMILES
NCCCCCCCC(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White

Description

8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text