Thermo Scientific Chemicals

Copper(II) phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17360.0E
2500 g, Each
Thermo Scientific Chemicals

Copper(II) phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A17360.0E
2500 g, Each
Quantity
Catalog number: A17360.0E
also known as A17360-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7798-23-4
IUPAC Name
tricopper(2+) diphosphate
Molecular Formula
Cu3O8P2
InChI Key
GQDHEYWVLBJKBA-UHFFFAOYSA-H
SMILES
[Cu++].[Cu++].[Cu++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Appearance (Color)
Pale green to green or pale blue to blue
Assay (Iodometric Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text