Copper(II) phosphate, 98%, Thermo Scientific Chemicals
Copper(II) phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) phosphate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A17360.36
also known as A17360-36
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
Add to cart
Copper(II) phosphate, 98%, Thermo Scientific Chemicals
Catalog numberA17360.36
Price (USD)/ Each
183.65
Online exclusive
204.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7798-23-4
IUPAC Nametricopper(2+) diphosphate
Molecular FormulaCu3O8P2
InChI KeyGQDHEYWVLBJKBA-UHFFFAOYSA-H
SMILES[Cu++].[Cu++].[Cu++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale green to green or pale blue to blue
Assay (Iodometric Titration)≥97.5 to ≤102.5%
FormPowder
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) phosphate is used as organic catalyst, fertilizer, emulsifier, corrosion inhibitor for phosphoric acid, metal protectant, and animal feed additive.

Solubility
Insoluble with water and ethanol. Soluble in ammonia, ammonium hydroxide, acetone.

Notes
Hygroscopic. Incompatible with strong bases, metals, alkali metals and powdered metals.
RUO – Research Use Only

General References:

  1. Zhang, Q.; Zhang, Z.; Teng, J.; Huang, H.; Peng, Q.; Jiao, T.; Hou, L.; Li, B. Highly Efficient Phosphate Sequestration in Aqueous Solutions Using Nanomagnesium Hydroxide Modified Polystyrene Materials. Ind. Eng. Chem. Res. 2015, 54 (11), 2940-2949.
  2. Zhao, J.; Tran, P. D.; Chen, Y.; Loo, J. S. C.; Barber, J.; Xu, Z. J. Achieving High Electrocatalytic Efficiency on Copper: A Low-Cost Alternative to Platinum for Hydrogen Generation in Water. J. Org. Chem. 2015, 5 (7), 4115-4120.