(-)-Diisopropyl D-tartrate, 98%, Thermo Scientific Chemicals
(-)-Diisopropyl D-tartrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(-)-Diisopropyl D-tartrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A17362.22
also known as A17362-22
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
(-)-Diisopropyl D-tartrate, 98%, Thermo Scientific Chemicals
Catalog numberA17362.22
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS62961-64-2
IUPAC Name1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC10H18O6
InChI KeyXEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILESCC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Optical Rotation-15.5 ± 1.0? (neat)
Assay (GC)≥97.5%
Refractive Index1.4375-1.4415 @ 20?C
FormLiquid or viscous liquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only