2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%, Thermo Scientific Chemicals
2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A17369.14
also known as A17369-14
Price (USD)/ Each
48.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
48.00
Add to cart
2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%, Thermo Scientific Chemicals
Catalog numberA17369.14
Price (USD)/ Each
48.00
-
Add to cart
Chemical Identifiers
CAS91-04-3
IUPAC Name2,6-bis(hydroxymethyl)-4-methylphenol
Molecular FormulaC9H12O3
InChI KeyKUMMBDBTERQYCG-UHFFFAOYSA-N
SMILESCC1=CC(CO)=C(O)C(CO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)> 88.0%
Appearance (Color)Cream or orange to pale brown
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only