Thermo Scientific Chemicals

tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A17373.36
500 g, Each
Thermo Scientific Chemicals

tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A17373.36
500 g, Each
Quantity
Catalog number: A17373.36
also known as A17373-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-45-9
IUPAC Name
tert-butyl benzenecarboperoxoate
Molecular Formula
C11H14O3
InChI Key
GJBRNHKUVLOCEB-UHFFFAOYSA-N
SMILES
CC(C)(C)OOC(=O)C1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Assay (Titration)
≥97.5 to ≤102.5%
Refractive Index
1.4970-1.5010 @ 20?C
Appearance (Color)
Clear, colorless to yellow

Description

tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

Solubility
Soluble in ether, alcohol, ester, and ketones. Insoluble in water. 

Notes
 Stored and transported as a mixture with inert solids and as a solvent slurry.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text