Thermo Scientific Chemicals

Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%, Thermo Scientific Chemicals

Catalog number: A17381.06
5 g, Each
Thermo Scientific Chemicals

Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%, Thermo Scientific Chemicals

Catalog number: A17381.06
5 g, Each
Quantity
Catalog number: A17381.06
also known as A17381-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3229-73-0
IUPAC Name
1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
Molecular Formula
C9H16N2O2
InChI Key
GMXOTWPDUBXCMS-UHFFFAOYSA-N
SMILES
CC1(C)C=C(C(N)=O)C(C)(C)N1O
Melting Point (clear melt)
197.0-204.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Yellow
Assay (GC)
≥98.5%

Description

It is employed as a highly reactive spin-label.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a highly reactive spin-label.

Solubility
Soluble in acetone, chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text