Thermo Scientific Chemicals

3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals

Catalog number: A17386.14
25 g, Each
Thermo Scientific Chemicals

3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals

Catalog number: A17386.14
25 g, Each
Quantity
Catalog number: A17386.14
also known as A17386-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-29-0
IUPAC Name
3-(4-chlorophenoxy)propane-1,2-diol
Molecular Formula
C9H11ClO3
InChI Key
MXOAEAUPQDYUQM-UHFFFAOYNA-N
SMILES
OCC(O)COC1=CC=C(Cl)C=C1
Appearance (Color)
White
Melting Point
76.5-82.5?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

Solubility
Slightly soluble in water.

Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text