Terbium(III) acetate hydrate, 99.99%, Thermo Scientific Chemicals
Terbium(III) acetate hydrate, 99.99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Terbium(III) acetate hydrate, 99.99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17388.06
also known as A17388-06
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
Add to cart
Terbium(III) acetate hydrate, 99.99%, Thermo Scientific Chemicals
Catalog numberA17388.06
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS100587-92-6
IUPAC Nameterbium(3+) triacetate
Molecular FormulaC6H9O6Tb
InChI KeyJQBILSNVGUAPMM-UHFFFAOYSA-K
SMILES[Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99.99%
FormCrystals or powder or crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only