Thermo Scientific Chemicals

O-Benzylhydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A17393.06
5 g, Each
Thermo Scientific Chemicals

O-Benzylhydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A17393.06
5 g, Each
Quantity
Catalog number: A17393.06
also known as A17393-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2687-43-6
IUPAC Name
hydrogen O-benzylhydroxylamine chloride
Molecular Formula
C7H10ClNO
InChI Key
HYDZPXNVHXJHBG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NOCC1=CC=CC=C1
Form
Crystals or powder or crystalline powder or flakes
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Water Content (Karl Fischer Titration)
≤0.5%
Assay (Titration ex Chloride)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Identification (FTIR)
Conforms (non-U.S. sourced material)

Description

For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

Solubility
Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text