Thermo Scientific Chemicals

Potassium trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A17398.14
25 g, Each
Thermo Scientific Chemicals

Potassium trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog number: A17398.14
25 g, Each
Quantity
Catalog number: A17398.14
also known as A17398-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2923-16-2
Form
Crystals or powder or crystalline powder or lumps
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤2.0%
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
138-145?C

Description

It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent for preparation of aryl trifluoromethyl sulfides, by heating with the diaryl disulfide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text