Ammonium hydrogen phosphate, 98%, Thermo Scientific Chemicals
Ammonium hydrogen phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydrogen phosphate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 g
Catalog number A17416.36
also known as A17416-36
Price (USD)/ Each
46.65
Online exclusive
51.60 
Save 4.95 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
51.60 
Save 4.95 (10%)
Add to cart
Ammonium hydrogen phosphate, 98%, Thermo Scientific Chemicals
Catalog numberA17416.36
Price (USD)/ Each
46.65
Online exclusive
51.60 
Save 4.95 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (US-sourced material)
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (UK-sourced material)
Identification (FTIR)Conforms (UK-sourced material)
FormCrystals or powder or crystalline powder or fused solid
View more
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.

Solubility
Soluble in w
RUO – Research Use Only

General References:

  1. PV Dharani Aiyer. Effect of C:N ratio on alpha amylase production by Bacillus licheniformis SPT 27. African Journal of Biotechnology. 2004, 3 (10),519-522.
  2. R.P. Rao.; D.J. Devine. RE-activated lanthanide phosphate phosphors for PDP applications. J. Lumin.. 2000, 87-891260-1263.