It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
RUO – Research Use Only
General References:
- J.Michael Hollas, et al. The C(1)-C(α) torsional potential function of 2-fluorostyrene from S1-S0 fluorescence in a supersonic jet.Chem. Phys. Lett.,1989,154(1), 14-19.
- Rosa M. Villamanan, et al. On the planarity of 2-fluorostyrene.J. Am. Chem. Soc.,1989,111(17), 6487-6491.