Thermo Scientific Chemicals

2-Fluorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A17425.22
100 g, Each
Thermo Scientific Chemicals

2-Fluorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A17425.22
100 g, Each
Quantity
Catalog number: A17425.22
also known as A17425-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
394-46-7
IUPAC Name
1-ethenyl-2-fluorobenzene
Molecular Formula
C8H7F
InChI Key
YNQXOOPPJWSXMW-UHFFFAOYSA-N
SMILES
FC1=CC=CC=C1C=C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Refractive Index
1.5180-1.5220 @ 20?C
Assay (GC)
≥97.5%

Description

It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text