It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.
Solubility
Solubility in hot water is almost transparent.
Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only
General References:
- Christopher E. Dahm, et al. Electrochemical and spectroscopic characterization of anodically formed nickel salen polymer films on glassy carbon, platinum, and optically transparent tin oxide electrodes in acetonitrile containing tetramethylammonium tetrafluoroborate.J. Electroanal. Chem.,1996,410(2), 163-171.
- Parichatr Vanalabhpatana, et al. Catalytic Reduction of 1,6-Dihalohexanes by Nickel(I) Salen Electrogenerated at Glassy Carbon Cathodes in Dimethylformamide.J. Electrochem. Soc.,2005,152(7), 222-E229.